Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylthiazolidine, 98%

Catalog No. aab2253714
Click to view available options
Quantity:
25 g

CAS: 24050-16-6 | C4H9NS | 103.183 g/mol

2-Methylthiazolidine is used as sources of low molecular-weight thiol compounds both in EATC and mouse liver homogenate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylthiazolidine is used as sources of low molecular-weight thiol compounds both in EATC and mouse liver homogenate.

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24050-16-6
Molecular Formula C4H9NS
Molecular Weight (g/mol) 103.183
MDL Number MFCD00053110
InChI Key DQMLFUMEBNHPPB-UHFFFAOYSA-N
PubChem CID 32196
IUPAC Name 2-methyl-1,3-thiazolidine
SMILES CC1NCCS1

Specifications

Boiling Point 76°C to 78°C (36 mmHg)
Flash Point 35°C (95°F)
Refractive Index 1.526
Quantity 25 g
UN Number UN1993
Synonym 2-methylthiazolidine, thiazolidine, 2-methyl, methyl-2 thiazolidine, methyl-2 thiazolidine french, 2-methyltetrahydrothiazole, acmc-20anpm, thiazolidine,2-methyl, 4-27-00-00661 beilstein handbook reference, +/--2-methylthiazolidine, #
Solubility Information Slightly soluble in water
Formula Weight 103.18
Percent Purity 99%
Chemical Name or Material 2-Methylthiazolidine
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECSNumber XJ6061000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.