Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4(1H)-one 98.0+%, TCI America™
SDP

Catalog No. M28511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28511G 1 g
M28515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28511G Supplier TCI America Supplier No. M28511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16097-62-4
Molecular Formula C6H5F3N2OS
Molecular Weight (g/mol) 210.174
MDL Number MFCD00153191
InChI Key BRLPDRZKOMZVOY-UHFFFAOYSA-N
Synonym 2-methylthio-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methylthio-6-trifluoromethyl pyrimidine, 2-methylsulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-methylsulfanyl-6-trifluoromethyl-4-pyrimidinol, methylsulfanyltrifluoromethylpyrimidinol, 2-methylsulfanyl-6-trifluoromethyl-1h-pyrimidin-4-one, 4-hydroxy-2-methylthio-6-trifluoromethyl-pyrimidine, pubchem16996, 2-methylthio-4-trifluoromethyl-6-pyrimidinol, 2-methylthio-6-trifluoromethylpyrimidin-4-ol
PubChem CID 248062
IUPAC Name 2-methylsulfanyl-6-(trifluoromethyl)-1H-pyrimidin-4-one
SMILES CSC1=NC(=O)C=C(N1)C(F)(F)F

Specifications

Melting Point 180°C
Color White
Quantity 1 g
Formula Weight 210.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4(1H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.