Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)benzoic acid, 98+%

Catalog No. aaa1037906
Click to view available options
Quantity:
5 g
25 g

CAS: 3724-10-5 | C8H8O2S | 168.21 g/mol

2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

Solubility
Soluble in Methanol.

Notes
Incompatible with strong oxidizing agents, acids, bases, amines. Store in a cool, dry place. Store in a tightly closed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3724-10-5
MDL Number MFCD00029934
ChEBI CHEBI:4657

Specifications

Melting Point 166°C to 168°C
Molecular Formula C21H29N3O
Quantity 5 g
Synonym disopyramide, dicorantil, rythmodan, disopiramida, ritmodan, disopyramidum, searle 703, isorythm, lispine, disopyramidum inn-latin
Solubility Information Soluble in Methanol.
InChI Key UVTNFZQICZKOEM-UHFFFAOYNA-N
SMILES CC(C)N(CCC(C1=CC=CC=C1)(C2=CC=CC=N2)C(=O)N)C(C)C
IUPAC Name 4-[di(propan-2-yl)amino]-2-phenyl-2-pyridin-2-ylbutanamide
Molecular Weight (g/mol) 339.48
PubChem CID 3114
Formula Weight 168.21
Percent Purity ≥98%
Chemical Name or Material 2- (Methylthio)benzoic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.