Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)ethylamine, 95%

Catalog No. p-7026749
Click to view available options
Quantity:
1 g
5 g

CAS: 18542-42-2 | C3H9NS | 91.172 g/mol

2-(Methylthio)ethylamine is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)ethylamine is a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents, carbon dioxide. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18542-42-2
Molecular Formula C3H9NS
Molecular Weight (g/mol) 91.172
MDL Number MFCD00014825
InChI Key CYWGSFFHHMQKET-UHFFFAOYSA-N
Synonym 2-methylthio ethylamine, 2-methylthio ethanamine, 2-aminoethyl methyl sulfide, 2-methylsulfanyl ethanamine, ethanamine, 2-methylthio, 2-methylsulfanyl ethan-1-amine, 2-methylthio ethyl amine, ethylamine, 2-methylthio, s-methylcysteamine
PubChem CID 87697
IUPAC Name 2-methylsulfanylethanamine
SMILES CSCCN

Specifications

Density 0.974
Boiling Point 146°C to 148°C
Flash Point 36°C (96°F)
Odor Amine-like
Refractive Index 1.4965
Linear Formula CH3S(CH2)2NH2
Quantity 1 g
UN Number UN2734
Beilstein 1731099
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 91.18
Percent Purity 95%
Chemical Name or Material 2-(Methylthio)ethylamine
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 242-412-5
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.