Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Methylthio)naphthalene, 98%

Click to view available options
Quantity:
1g
5g
25g
7433-79-6
C11H10S
174.261
MFCD00051615
SSKUUDUKJMIOKQ-UHFFFAOYSA-N
2-methylthio naphthalene,2-methylmercaptonaphthalene,2-methylsulfanyl naphthalene,2-methylthionaphthalene,methyl2-naphthylsulfide,methyl 2-naphthyl sulfide,naphth-2-yl methyl thioether,naphthalene,2-methylthio,methyl naphthalen-2-yl sulfane
139022
2-methylsulfanylnaphthalene
CSC1=CC2=CC=CC=C2C=C1
7433-79-6
C11H10S
174.261
MFCD00051615
SSKUUDUKJMIOKQ-UHFFFAOYSA-N
2-methylthio naphthalene,2-methylmercaptonaphthalene,2-methylsulfanyl naphthalene,2-methylthionaphthalene,methyl2-naphthylsulfide,methyl 2-naphthyl sulfide,naphth-2-yl methyl thioether,naphthalene,2-methylthio,methyl naphthalen-2-yl sulfane
139022
2-methylsulfanylnaphthalene
CSC1=CC2=CC=CC=C2C=C1
Applications
2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Melting Point 62°C to 63°C
Quantity 5g
Odor Stench
Beilstein 1859409
Formula Weight 174.27
Percent Purity 98%
Chemical Name or Material 2-(Methylthio)naphthalene