Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 2-Methylthioadenosine diphosphate trisodium salt
GREENER_CHOICESDP

Catalog No. 162410 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-241-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 16-241-0 Supplier Tocris Bioscience™ Supplier No. 1624/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent agonist at P2Y1, P2Y12 and P2Y13

Potent purinergic agonist displaying selectivity for P2Y1, P2Y12 and P2Y13 receptors (pEC50 = 8.29 and 9.05 for P2Y1 and P2Y12, EC50 = 19 nM for P2Y13). Induces aggregation of, and inhibits cAMP accumulation in, platelets in vitro.

Chemical Identifiers

CAS 475193-31-8
Molecular Formula C11H17N5Na3O10P2S

Specifications

Quantity 10 mg
Formula Weight 539.24
Purity >98%
Molecular Weight (g/mol) 542.259
Chemical Name or Material 2-Methylthioadenosine diphosphate trisodium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.