Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 2-Methylthioadenosine triphosphate tetrasodium salt
GREENER_CHOICESDP

Catalog No. 106210 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-621-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-621-0 Supplier R&D Systems™ Supplier No. 1062/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-selective P2 agonist

P2 purinoceptor agonist.

Chemical Identifiers

CAS 100020-57-3
Molecular Formula C11H14N5Na4O13P3S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 641.2
Purity >98%
Molecular Weight (g/mol) 641.195
Chemical Name or Material 2-Methylthioadenosine triphosphate tetrasodium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.