Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
β2 Microglobulin Mouse anti-Mouse, BUV563, Clone: TLD-3H12B, BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 748958
Description
β2 Microglobulin Monoclonal antibody specifically detects β2 Microglobulin in Mouse samples. It is validated for Flow Cytometry
Specifications
β2 Microglobulin | |
Monoclonal | |
Brilliant Ultraviolet 563 | |
B2m; Beta-2-microglobulin | |
Cultured Lewis rat microglial cells | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
TLD-3H12B | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction