Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Morpholinoaniline 98.0+%, TCI America™
SDP

Catalog No. M267125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M267125G 25 g
M26715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M267125G Supplier TCI America Supplier No. M267125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5585-33-1
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00047408
InChI Key QKWLVAYDAHQMLG-UHFFFAOYSA-N
Synonym 2-morpholinoaniline, 2-morpholin-4-yl aniline, 2-morpholin-4-yl-phenylamine, 2-4-morpholino aniline, 4-2-aminophenyl morpholine, 2-4-morpholinyl aniline, 2-morpholin-4-ylphenylamine, enamine_000782, acmc-1ap2s
PubChem CID 735756
IUPAC Name 2-morpholin-4-ylaniline
SMILES C1COCCN1C2=CC=CC=C2N

Specifications

Melting Point 96°C
Color White-Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 178.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Morpholinoaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.