Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Butoxyethanol, 99%

Catalog No. AAA179760D
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA179760D 10,000 mL
AAA17976AP 500 mL
AAA179760F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA179760D Supplier Thermo Scientific Chemicals Supplier No. A179760D
Add to Cart

CAS: 111-76-2 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Used in industry for degreasing and detergent applications.2-n-Butoxyethanol is used in the industry for degreasing and detergent applications. It acts as a surfactant in domestic and industrial products. It is an active component of fracturing fluids, drilling stabilizers and oil slick dispersants. It is also used as a solvent for paints, surface coatings, cleaning products and inks. Further, it is employed as a food additive, antimicrobial agent, defoamer, stabilizer and adhesive. In addition to this, it is used as a whiteboard cleaner, liquid soap, cosmetic, dry cleaning solution, lacquer, varnishe and latex paint.

Solubility
Miscible with water, ethyl alcohol, ethyl ether, acetone, n-heptane benzene, mineral oil, aromatic paraffin and halogenated hydrocarbons. Slightly miscible with carbon tetrachloride.

Notes
Excellent Hydrophilic-lipophilic balance (HLB).Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, metals, aluminum and alkalies.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00002884
InChI Key POAOYUHQDCAZBD-UHFFFAOYSA-N
Synonym butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb
PubChem CID 8133
ChEBI CHEBI:63921
IUPAC Name 2-butoxyethanol
SMILES CCCCOCCO

Specifications

Melting Point -79°C
Density 0.902
Boiling Point 171°C to 173°C
Flash Point 64°C (147°F)
Odor Mild
Assay Percent Range 0.98
Refractive Index 1.419
Linear Formula CH3(CH2)3OCH2CH2OH
Quantity 10,000 mL
UN Number UN2810
Beilstein 1732511
Merck Index 14,1559
Solubility Information Miscible with water,ethyl alcohol,ethyl ether,acetone,n-heptane benzene,mineral oil,aromatic paraffin and halogenated hydrocarbons. Slightly miscible with carbon tetrachloride.
Formula Weight 118.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-n-Butoxyethanol
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 203-905-0
RTECSNumber KJ8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.