Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(N-Ethylperfluorooctanesulfonamido)ethylacrylate, Stabilized, Thermo Scientific™

Catalog No. AC410330250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC410330250 25 g
AC410331000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC410330250 Supplier Thermo Scientific Chemicals Supplier No. 410330250
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 423-82-5
Molecular Formula C15H12F17NO4S
Molecular Weight (g/mol) 625.297
InChI Key ZAZJGBCGMUKZEL-UHFFFAOYSA-N
Synonym 2-n-ethylperfluorooctanesulfonamido ethyl acrylate, unii-7l81f9qvy8, 2-ethyl heptadecafluorooctyl sulfonyl amino ethyl acrylate, 2-ethyl heptadecafluorooctyl sulphonyl amino ethyl acrylate, 2-ethyl 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctylsulfonyl amino ethyl prop-2-enoate, 2-propenoic acid, 2-ethyl 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl sulfonyl amino ethyl ester
PubChem CID 67921
IUPAC Name 2-[ethyl(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctylsulfonyl)amino]ethyl prop-2-enoate
SMILES CCN(CCOC(=O)C=C)S(=O)(=O)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Linear Formula CF3(CF2)7SO2N(CH2CH3)CH2CH2O2CCHCH2
Quantity 25 g
Formula Weight 625.29
Chemical Name or Material 2-(N-Ethylperfluorooctanesulfonamido)ethylacrylate
EINECSNumber 207-031-0
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.