Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthalenemethanol, 98%

Catalog No. p-3758537
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 1592-38-7 | C11H10O | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1592-38-7
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004124
InChI Key MFGWMAAZYZSWMY-UHFFFAOYSA-N
Synonym 2-naphthalenemethanol, 2-naphthylmethanol, 2-hydroxymethylnaphthalene, 2-naphthalene methanol, 2-naphthyl methanol, naphthalen-2-yl-methanol, naphthalen-2-yl methanol, 2-naphthylmethan-1-ol, 2-naphthylcarbinol, 2na
PubChem CID 74128
ChEBI CHEBI:27615
IUPAC Name naphthalen-2-ylmethanol
SMILES C1=CC=C2C=C(C=CC2=C1)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 79°C to 83°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH2OH
Quantity 1 g
Formula Weight 158.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthalenemethanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.