Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthalenemethanol 98.0+%, TCI America™
SDP

Catalog No. N088125G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1592-38-7
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004124
InChI Key MFGWMAAZYZSWMY-UHFFFAOYSA-N
Synonym 2-naphthalenemethanol, 2-naphthylmethanol, 2-hydroxymethylnaphthalene, 2-naphthalene methanol, 2-naphthyl methanol, naphthalen-2-yl-methanol, naphthalen-2-yl methanol, 2-naphthylmethan-1-ol, 2-naphthylcarbinol, 2na
PubChem CID 74128
ChEBI CHEBI:27615
IUPAC Name naphthalen-2-ylmethanol
SMILES C1=CC=C2C=C(C=CC2=C1)CO

Specifications

Melting Point 80°C
Color White-Yellow
Boiling Point 178°C
Quantity 25 g
Formula Weight 158.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthalenemethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.