Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthonitrile, 97%

Catalog No. p-171152
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC346310050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC346310050 Supplier Thermo Scientific Chemicals Supplier No. 346310050

CAS: 613-46-7 | C11H7N | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-46-7
Molecular Formula C11H7N
Molecular Weight (g/mol) 153.18
MDL Number MFCD00016807
InChI Key AZKDTTQQTKDXLH-UHFFFAOYSA-N
Synonym 2-naphthonitrile, 2-cyanonaphthalene, 2-naphthalenecarbonitrile, 2-cyanonaphthylene, 2-naphthalenenitrile, beta-cyanonaphthalene, beta-naphthonitrile, .beta.-naphthonitrile, naphthalene-2-carbonitrile, 2-naphthylcyanide
PubChem CID 11944
IUPAC Name naphthalene-2-carbonitrile
SMILES C1=CC=C2C=C(C=CC2=C1)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 63°C to 68°C
Color White to Beige
Boiling Point 156°C to 158°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 153.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthonitrile, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
EINECSNumber 210-344-5
RTECSNumber QL5976350
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.