Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthoyl chloride, 98%, Acros Organics

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
2243-83-6
C11H7ClO
190.63
MFCD00004093
XNLBCXGRQWUJLU-UHFFFAOYSA-N
2-naphthoyl chloride,2-naphthalenecarbonyl chloride,2-naphthoic chloride,2-naphthoylchloride,beta-naphthoyl chloride,2-napthoyl chloride,2-chlorocarbonyl naphthalene,.beta.-naphthoyl chloride,.beta.-naphthalenecarbonyl chloride,beta-naphthalenecarbonyl chloride
75246
naphthalene-2-carbonyl chloride
C1=CC=C2C=C(C=CC2=C1)C(=O)Cl
2243-83-6
C11H7ClO
190.63
MFCD00004093
XNLBCXGRQWUJLU-UHFFFAOYSA-N
2-naphthoyl chloride,2-naphthalenecarbonyl chloride,2-naphthoic chloride,2-naphthoylchloride,beta-naphthoyl chloride,2-napthoyl chloride,2-chlorocarbonyl naphthalene,.beta.-naphthoyl chloride,.beta.-naphthalenecarbonyl chloride,beta-naphthalenecarbonyl chloride
75246
naphthalene-2-carbonyl chloride
C1=CC=C2C=C(C=CC2=C1)C(=O)Cl
Assay Percent Range 97.5% min. (GC)
Linear Formula C10H7COCl
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in aceton
Formula Weight 190.63
Physical Form Crystalline Chunks
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Solubility Solubility in water: decomposes. Other solubilities: soluble in aceton
Boiling Point 156.0°C to 158.0°C (7.0mmHg)
Color Brown to Gray
Melting Point 49.0°C to 54.0°C
Quantity 50g
Flash Point >110°C
Chemical Name or Material 2-Naphthoyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 218-822-5