Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthyl isocyanate, 97%, Thermo Scientific™

Catalog No. AC319440010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 2243-54-1
Molecular Formula C11H7NO
Molecular Weight (g/mol) 169.19
MDL Number MFCD00014319
InChI Key XIXJQNFTNSQTBT-UHFFFAOYSA-N
Synonym 2-naphthyl isocyanate, 2-naphthylisocyanate, naphthalene, 2-isocyanato, beta-naphthylisocyanate, unii-9ze6124voi, naphthalene,2-isocyanato, naphthalen-2-isocyanate, 2-naphthalenyl isocyanate, 2-isocyanato-naphthalene, acmc-1cr5l
PubChem CID 16718
IUPAC Name 2-isocyanatonaphthalene
SMILES C1=CC=C2C=C(C=CC2=C1)N=C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 52.0°C to 57.0°C
Color White
Boiling Point 83.0°C (0.2 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 169.19
Percent Purity 97%
Physical Form Chunks
Chemical Name or Material 2-Naphthyl isocyanate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
May cause allergy or asthma sympto
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comforta
EINECSNumber 218-815-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.