Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthyl sulfate potassium salt, 98%

Catalog No. AA4406103
Click to view available options
Quantity:
1 g

CAS: 1733-89-7 | C10H8KO4S | 263.328 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1733-89-7
Molecular Formula C10H8KO4S
Molecular Weight (g/mol) 263.328
MDL Number MFCD00004059
InChI Key KPQVGLIXPMOHMI-UHFFFAOYSA-N
Synonym 2-naphthyl hydrogen sulfate
PubChem CID 24186395
IUPAC Name naphthalen-2-yl hydrogen sulfate;potassium
SMILES C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)O.[K]

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 262.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Naphthyl sulfate potassium salt
EINECSNumber 217-072-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.