Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzaldehyde, 97%, Thermo Scientific™

Catalog No. sb01264ea
Click to view available options
Quantity:
10 g
50 g
100 g

Chemical Identifiers

CAS 552-89-6
Molecular Formula C7H5NO3
Molecular Weight (g/mol) 151.121
MDL Number MFCD00007132
InChI Key CMWKITSNTDAEDT-UHFFFAOYSA-N
Synonym o-nitrobenzaldehyde, benzaldehyde, 2-nitro, nitrobenzaldehyde, benzaldehyde, o-nitro, 2-nitro-benzaldehyde, 2-formylnitrobenzene, ccris 2322, unii-48b18q9b8e, benzaldehyde, nitro, ortho-nitrobenzaldehyde
PubChem CID 11101
ChEBI CHEBI:66927
IUPAC Name 2-nitrobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)[N+](=O)[O-]

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula O2NC6H4CHO
Quantity 10 g
Formula Weight 151.12
Percent Purity 97%
Chemical Name or Material 2-Nitrobenzaldehyde
Hazard Category Warning
Hazard Statement GHS P Statement Harmful to aquatic life with long lasting effects. Harmful if swallowed. Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously w
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.