Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzaldehyde, 98+%

Catalog No. AAA1150122
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 552-89-6 | C7H5NO3 | 151.121 g/mol

2-Nitrobenzaldehyde is used as a starting material in synthetic chemistry. It is used as an intermediate during the synthesis of Indigo dye. It is involved in the preparation of colorants such as Indigo carmine. Further, it serves as a useful photoremovable protecting group as well as utilized in the preparation of o-nitrophenylethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzaldehyde is used as a starting material in synthetic chemistry. It is used as an intermediate during the synthesis of Indigo dye. It is involved in the preparation of colorants such as Indigo carmine. Further, it serves as a useful photoremovable protecting group as well as utilized in the preparation of o-nitrophenylethylene glycol.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-89-6
Molecular Formula C7H5NO3
Molecular Weight (g/mol) 151.121
MDL Number MFCD00007132
InChI Key CMWKITSNTDAEDT-UHFFFAOYSA-N
Synonym o-nitrobenzaldehyde, benzaldehyde, 2-nitro, nitrobenzaldehyde, benzaldehyde, o-nitro, 2-nitro-benzaldehyde, 2-formylnitrobenzene, ccris 2322, unii-48b18q9b8e, benzaldehyde, nitro, ortho-nitrobenzaldehyde
PubChem CID 11101
ChEBI CHEBI:66927
IUPAC Name 2-nitrobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)[N+](=O)[O-]

Specifications

Melting Point 43°C to 46°C
Boiling Point 153°C (23 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 742624
Sensitivity Air and light sensitive
Merck Index 14,6587
Solubility Information Insoluble in water.
Formula Weight 151.12
Percent Purity ≥98%
Chemical Name or Material 2-Nitrobenzaldehyde
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-025-3
RTECSNumber CU7300000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.