Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzenesulfonamide, 97+%

Catalog No. AAA1568914
Click to view available options
Quantity:
5 g
10 g
25 g
100 g

CAS: 5455-59-4 | C6H6N2O4S | 202.18 g/mol

It is applied as a reagent for synthesis of medium-ring amines via N-alkylation, e.g. with ω-bromo alkanols under Mitsunobu conditions (DEAD/PPh3), and cyclization of the resulting ω-bromo alkylsulfonamides (Cs2CO3/Bu4NI). It is a carbonic anhydrase inhibitor. It is employed as a reactant involved in synthesis of cyclic nitrogen compounds via intramolecular hydroamination, pentacyclic lycopodium alkaloid huperzine-Q2, pyrrolidines, intermolecular C-H insertion reactions and reactant involved in intermolecular amination of allyl alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a reagent for synthesis of medium-ring amines via N-alkylation, e.g. with ω-bromo alkanols under Mitsunobu conditions (DEAD/PPh3), and cyclization of the resulting ω-bromo alkylsulfonamides (Cs2CO3/Bu4NI). It is a carbonic anhydrase inhibitor. It is employed as a reactant involved in synthesis of cyclic nitrogen compounds via intramolecular hydroamination, pentacyclic lycopodium alkaloid huperzine-Q2, pyrrolidines, intermolecular C-H insertion reactions and reactant involved in intermolecular amination of allyl alcohols.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5455-59-4
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.18
MDL Number MFCD00009807
InChI Key GNDKYAWHEKZHPJ-UHFFFAOYSA-N
Synonym 2-nitrobenzene-1-sulfonamide, 2-nitrobenzenesulphonamide, 2-nitro-benzenesulfonamide, nitrobenzenesulfonamide, 2-nitrophenylsulfonamide, 2-sulphamoylnitrobenzene, acmc-209lhg, 2-nitrobenzene-sulfonamide, 2-nitro benzene sulphonamide, ksc490i0n
PubChem CID 138510
IUPAC Name 2-nitrobenzenesulfonamide
SMILES NS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 188°C to 194°C
Quantity 25 g
Beilstein 2214215
Solubility Information Slightly soluble in water.
Formula Weight 202.19
Percent Purity ≥98%
Chemical Name or Material 2-Nitrobenzenesulfonamide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.