Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzyl chloride, 98%

Catalog No. p-172846
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle

CAS: 612-23-7 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-23-7
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007185
InChI Key BXCBUWKTXLWPSB-UHFFFAOYSA-N
Synonym 2-nitrobenzyl chloride, 1-chloromethyl-2-nitrobenzene, o-nitrobenzyl chloride, alpha-chloro-2-nitrotoluene, benzene, 1-chloromethyl-2-nitro, 2-nitrobenzylchloride, unii-hb8u484npm, alpha-chloro-o-nitrotoluene, ccris 2323, toluene, alpha-chloro-o-nitro
PubChem CID 11921
IUPAC Name 1-(chloromethyl)-2-nitrobenzene
SMILES C1=CC=C(C(=C1)CCl)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 44°C to 49°C
Color Tan to Yellow
Boiling Point 127°C to 133°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4CH2Cl
Quantity 50 g
Beilstein 05, 327
Solubility Information Solubility in water: insoluble in water
Formula Weight 171.58
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2-Nitrobenzyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 210-300-5
RTECSNumber XS9092000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.