Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitroimidazole, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
527-73-1
C3H3N3O2
113.08
MFCD00005185
YZEUHQHUFTYLPH-UHFFFAOYSA-N
2-nitroimidazole,azomycin,amicin,nitroimidazole,1h-imidazole, 2-nitro,imidazole, 2-nitro,2-nitroimidazol,2-nitro-imidazole,unii-k8e96xl55d,azomycin 2-nitroimidazole
10701
CHEBI:67135
2-nitro-1H-imidazole
C1=CN=C(N1)[N+](=O)[O-]
527-73-1
C3H3N3O2
113.08
MFCD00005185
YZEUHQHUFTYLPH-UHFFFAOYSA-N
2-nitroimidazole,azomycin,amicin,nitroimidazole,1h-imidazole, 2-nitro,imidazole, 2-nitro,2-nitroimidazol,2-nitro-imidazole,unii-k8e96xl55d,azomycin 2-nitroimidazole
10701
CHEBI:67135
2-nitro-1H-imidazole
C1=CN=C(N1)[N+](=O)[O-]
Melting Point 287°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color Green-Khaki to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 15, 914
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alkaline h2o- insoluble in acidic h2o,soluble in methanol,ethanol,acetone,ethyl,acetate and butyl acetate,practically insoluble in ether and chloroform
Formula Weight 113.08
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Nitroimidazole, 98%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye prot

GHS Signal Word: Danger

EINECSNumber : 208-425-5

RTECSNumber : NI7875000

TSCA : TSCA