Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc acetate dihydrate, 97%, specified according to the requirements of USP
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 237.0°C |
Quantity | 2.5 kg |
Assay Percent Range | 98.0 to 102% (Complexometry) |
Linear Formula | (CH3CO2)2Zn·2H2O |
Merck Index | 15,10327 |
Solubility Information | Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol |
Formula Weight | 219.5 |
Percent Purity | 97% |
Packaging | Plastic bottle |
Specific Gravity | 1.84 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful if swallowed. Very toxic to aquatic life. |
Precautionary Statement | GHS P Statement Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously wit |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction