Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitropropane 95.0+%, TCI America™
SDP

Catalog No. N0249100G
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 79-46-9
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00007397
InChI Key FGLBSLMDCBOPQK-UHFFFAOYSA-N
Synonym propane, 2-nitro, isonitropropane, dimethylnitromethane, nitroisopropane, sec-nitropropane, nipar s-20, beta-nitropropane, 2-np, nipar s-20 solvent, nipar s-30 solvent
PubChem CID 398
ChEBI CHEBI:16037
IUPAC Name 2-nitropropane
SMILES CC(C)[N+](=O)[O-]

Specifications

Melting Point -91°C
Color Yellow
Boiling Point 120°C
Quantity 100 g
UN Number 2608
Formula Weight 89.09
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Nitropropane
EINECSNumber (2)-0194
RTECSNumber TZ5250000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.