Learn More
2-Nitrotoluene, 99+%
Description
2-Nitrotoluene is used as an intermediate for the o-toludine synthesis, which finds application in the dyestuff industry. It is employed as a reagent in detecting and photometric determining oxidizing agent. It is also used in the determination of prussic acid content in the air. It acts as a precursor to prepare 2-amino-4-chlorotoluene, 2-amino-6-chlorotoluene, nitrotoluenesulfonic acids and toluene nitrosulfonic chlorides. It acts as a nitrogen supplement in the culture medium of pseudomonas sp. strain ClS1. It acts as a carbon and energy supplement in the culture medium of Pseudomonas sp. strain JS42.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Nitrotoluene is used as an intermediate for the o-toludine synthesis, which finds application in the dyestuff industry. It is employed as a reagent in detecting and photometric determining oxidizing agent. It is also used in the determination of prussic acid content in the air. It acts as a precursor to prepare 2-amino-4-chlorotoluene, 2-amino-6-chlorotoluene, nitrotoluenesulfonic acids and toluene nitrosulfonic chlorides. It acts as a nitrogen supplement in the culture medium of pseudomonas sp. strain ClS1. It acts as a carbon and energy supplement in the culture medium of Pseudomonas sp. strain JS42.
Solubility
Miscible with acetone, benzene, chloroform, diethyl ether, ethanol and petroleum ether. Immiscible with water.
Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases, sulfuric acid, reducing agents, hydrogen and sodium.
Specifications
Specifications
| Melting Point | -4°C |
| Density | 1.166 |
| Boiling Point | 224°C to 226°C |
| Flash Point | 106°C (222°F) |
| Refractive Index | 1.545 |
| Quantity | 100 g |
| UN Number | UN1664 |
| Beilstein | 1907580 |
| Merck Index | 14,6650 |
| Solubility Information | Miscible with acetone,benzene,chloroform,diethyl ether,ethanol and petroleum ether; Immiscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H302-H340-H350-H361f |
| Hazard Statement | GHS H Statement H340-H350-H361-H302 May cause genetic defects. May cause cancer. Suspected of damaging fertility or the unborn child. Harmful if swallowed. |
| Precautionary Statement | P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROTOLUENES, LIQUID |
| EINECSNumber | 201-853-3 |
| RTECSNumber | XT3150000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.