Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 2'-O-methylcytidine Recombinant Rabbit Monoclonal Antibody (8Q7I8)
GREENER_CHOICE

Catalog No. PIMA542763
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542763 Supplier Invitrogen™ Supplier No. MA542763
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: Cm conjuga ted to KLH.

2'-O-Methylcytidine is derivative of the nucleoside residue cytidine with a methyl group attached to the 2'-oxygen position.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 2'-O-methylcytidine
Applications Dot Blot, ELISA
Classification Recombinant Monoclonal
Clone 8Q7I8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Alias 2'-O-methylcytidine; methylcytidine
Host Species Rabbit
Immunogen Chemical compounds corresponding to 2'-O-methylcytidine/ Cm.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical, Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.