Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-O-Methylcytidine 98.0+%, TCI America™
SDP

Catalog No. M23171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23171G 1 g
M2317200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23171G Supplier TCI America Supplier No. M23171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2140-72-9
Molecular Formula C10H15N3O5
Molecular Weight (g/mol) 257.246
MDL Number MFCD00056067
InChI Key RFCQJGFZUQFYRF-ZOQUXTDFSA-N
PubChem CID 150971
ChEBI CHEBI:19228
IUPAC Name 4-amino-1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]pyrimidin-2-one
SMILES COC1C(C(OC1N2C=CC(=NC2=O)N)CO)O

Specifications

Melting Point 257°C
Color White
Quantity 1 g
Synonym 2'-o-methylcytidine, cytidine, 2'-o-methyl, 2'-o-methyl cytidine, o 2'-methylcytidine, 4-amino-1-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl pyrimidin-2 1h-one, 2'-o-methyl-cytidine, o-2-methylcytidine, o2'-methylcytidine, 2-o-methylcytidine, 2/'-o-methylcytidine
Formula Weight 257.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-O-Methylcytidine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.