Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Octen-4-one, 97%
Click to view available options
Quantity:
1 g
5 g
Description
Reactant in the rhodium-catalyzed asymmetric ring opening of oxabicyclic alkenes with organoboronic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant in the rhodium-catalyzed asymmetric ring opening of oxabicyclic alkenes with organoboronic acids.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Reactant in the rhodium-catalyzed asymmetric ring opening of oxabicyclic alkenes with organoboronic acids.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Density | 0.85 |
Boiling Point | 65°C (15 mmHg) |
Flash Point | 58°C (136°F) |
Refractive Index | 1.445 |
Quantity | 1 g |
UN Number | UN1224 |
Solubility Information | Insoluble in water. |
Formula Weight | 126.2 |
Percent Purity | 97% |
Chemical Name or Material | 2-Octen-4-one |
Safety and Handling
Hazard Category | H226-H315-H319 |
Hazard Statement | GHS H Statement H226 Flammable liquid and vapour. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S. |
EINECSNumber | 225-071-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction