Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Oxa-5-azaspiro[3.4]octane oxalate, 96%

Catalog No. aah57854md
Click to view available options
Quantity:
250 mg
500 mg

CAS: 1389264-18-9 | C8H13NO5 | 203.194 g/mol

Spiro forms of lactones and oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photochromic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Spiro forms of lactones and oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photochromic materials.

Solubility
Decomposes in water.

Notes
Keep container tightly closed. Keep container in a cool refrigerated, well-ventilated area. Moisture sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1389264-18-9
Molecular Formula C8H13NO5
Molecular Weight (g/mol) 203.194
MDL Number MFCD18325162
InChI Key JFOZNINEJYPQQK-UHFFFAOYSA-N
Synonym 2-oxa-5-azaspiro 3.4 octane oxalate, 2-oxa-5-aza-spiro 3.4 octane oxalate, 2-oxa-5-azaspiro 3.4 octane oxalate salt, 2-oxa-5-azaspiro 3.4 octane, hemioxalate salt, 2-oxa-5-azaspiro 3.4 octane; oxalic acid
PubChem CID 72207353
IUPAC Name 2-oxa-5-azaspiro[3.4]octane;oxalic acid
SMILES C1CC2(COC2)NC1.C(=O)(C(=O)O)O

Specifications

Quantity 250 mg
Solubility Information Decomposes in water.
Formula Weight 203.19
Percent Purity 96%
Chemical Name or Material 2-Oxa-5-azaspiro[3.4]octane oxalate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.