Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Oxazolidinone, 99%

Catalog No. p-7021982
Click to view available options
Quantity:
25 g
100 g
500 g

Catalog No. AAA1454314

Supplier: Thermo Scientific Chemicals A1454314

Only null left
Add to Cart

CAS: 497-25-6 | C3H5NO2 | 87.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Oxazolidinone is used as antimicrobials, in organisms it act as a protein synthesis inhibitors, targeting an early step involving the binding of N-formylmethionyl-tRNA to the ribosome. They are used for chiral synthesis. It has been used as an ethanolamine equivalent, also behaves as an aziridine equivalent as in the conversion of thiophenols or amines to their aminoethyl derivatives.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-25-6
Molecular Formula C3H5NO2
Molecular Weight (g/mol) 87.08
MDL Number MFCD00005268
InChI Key IZXIZTKNFFYFOF-UHFFFAOYSA-N
Synonym 2-oxazolidone, oxazolidin-2-one, 2-oxazolidinone, oxazolidinone, oxazolidone, oxazolidinones, unii-z4d49w92pp, carbamic acid, 2-hydroxyethyl-, gamma-lactone, wln: t5mvotj, carbamic acid, .gamma.-lactone
PubChem CID 73949
ChEBI CHEBI:1237
IUPAC Name 1,3-oxazolidin-2-one
SMILES O=C1NCCO1

Specifications

Melting Point 84°C to 89°C
Boiling Point 220°C (48 mmHg)
Quantity 25 g
Beilstein 106251
Solubility Information Soluble in water.
Formula Weight 87.08
Percent Purity 99%
Chemical Name or Material 2-Oxazolidinone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 207-840-9
RTECSNumber RQ2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.