Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Oxo-1-pyrrolidinyl)acetic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-7056994
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53934-76-2
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.142
MDL Number MFCD00962836
InChI Key JGPIWNNFLKDTSR-UHFFFAOYSA-N
Synonym 2-oxo-1-pyrrolidineacetic acid, 2-2-oxopyrrolidin-1-yl acetic acid, 2-oxopyrrolidin-1-yl acetic acid, 2-oxo-pyrrolidin-1-yl-acetic acid, 2-oxo-1-pyrrolidinyl acetic acid, 1-pyrrolidineacetic acid, 2-oxo, 2-2-oxopyrrolidinyl acetic acid, piracetam impuirty d, 2-pyrrolidone acetic acid, acmc-209lb1
PubChem CID 3146687
IUPAC Name 2-(2-oxopyrrolidin-1-yl)acetic acid
SMILES C1CC(=O)N(C1)CC(=O)O

Specifications

Quantity 1 g
Beilstein 4342
Formula Weight 143.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (2-Oxo-1-pyrrolidinyl)acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.