Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Oxoglutaric Acid 99.0+%, TCI America™
SDP

Catalog No. K0005100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K0005100G 100 g
K000525G 25 g
K0005500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K0005100G Supplier TCI America Supplier No. K0005100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 328-50-7
Molecular Formula C5H6O5
Molecular Weight (g/mol) 146.098
MDL Number MFCD00004165
InChI Key KPGXRSRHYNQIFN-UHFFFAOYSA-N
Synonym 2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid
PubChem CID 51
ChEBI CHEBI:30915
IUPAC Name 2-oxopentanedioic acid
SMILES C(CC(=O)O)C(=O)C(=O)O

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 100 g
Formula Weight 146.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Oxoglutaric Acid
EINECSNumber (9)-0467
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.