Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(p-Tolyl)pyridine 97.0+%, TCI America™
SDP

Catalog No. T122825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T122825G 25 g
T12285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T122825G Supplier TCI America Supplier No. T122825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4467-06-5
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006281
InChI Key KJNZQKYSNAQLEO-UHFFFAOYSA-N
Synonym 2-p-tolyl pyridine, 2-4-methylphenyl pyridine, 2-p-tolylpyridine, 2-4-tolyl pyridine, pyridine, 2-4-methylphenyl, pyridine,2-p-tolyl, 2-p-toluyl pyridine, pubchem16062, 4-2-pyridyl toluene
PubChem CID 78219
IUPAC Name 2-(4-methylphenyl)pyridine
SMILES CC1=CC=C(C=C1)C2=CC=CC=N2

Specifications

Boiling Point 157°C
Quantity 25 g
Formula Weight 169.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(p-Tolyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.