Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Pentanol

Catalog No. P8017500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Catalog No. Quantity
P8017500ML 500 mL
P80174L 4 L
P80171L 1 L
P8017100ML 100 mL
4 options

Catalog No. P8017500ML

Supplier: Sigma Aldrich P8017500ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-pentanol is a secondary alcohol that can be used as:
  • A hydrogen donor in the synthesis of methyl furan via catalytic transfer hydrogenation of furfural over a ruthenium/ruthenium oxide/carbon catalyst
  • A reagent in one-pot synthesis of brominated derivatives of 1,3-dioxolane
  • A solvent for the synthesis of organic molecules such as 2, 4-diaminopyrimidines and 4-aza-podophyllotoxins

Specifications

CAS 6032-29-7
Density 0.812 g/mL (at 25°C (literature))
Boiling Point 118°C to 119°C (lit.)
Molecular Formula C5H12O
Refractive Index n20/D 1.406 (literature)
Linear Formula CH3CH2CH2CH(OH)CH3
MDL Number MFCD00004579
Quantity 500 mL
Synonym (+/-)-2-Pentanol; sec-amyl alcohol; sec-Pentyl alcohol; Methyl propyl carbinol
Molecular Weight (g/mol) 88.15
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.