Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pentanone, 99+%, purified by redistillation, AcroSeal™

Catalog No. p-4812841
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 107-87-9 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-87-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
InChI Key XNLICIUVMPYHGG-UHFFFAOYSA-N
Synonym 2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone
PubChem CID 7895
ChEBI CHEBI:16472
IUPAC Name pentan-2-one
SMILES CCCC(=O)C

Specifications

Melting Point -78.0°C
Density 0.8090g/mL
Boiling Point 100.0°C to 103.0°C
Flash Point 7°C
Assay Percent Range 99% min. (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.3890 to 1.3910
Linear Formula CH3CO(CH2)CH3
Quantity 100 mL
Beilstein 01,676
Merck Index 15,6187
Specific Gravity 0.809
Solubility Information Solubility in water: 43g/L (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 0.5 mPa.s (20°C)
Formula Weight 86.13
Percent Purity 99+%
Chemical Name or Material 2-Pentanone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
EINECSNumber 203-528-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.