Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pentanone, 99%, pure, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
107-87-9
C5H10O
86.13
XNLICIUVMPYHGG-UHFFFAOYSA-N
2-pentanone,methyl propyl ketone,ethyl acetone,pentanone,ethylacetone,propyl methyl ketone,metylopropyloketon,methyl n-propyl ketone,methylpropyl ketone,methyl-propyl-cetone
7895
CHEBI:16472
pentan-2-one
CCCC(=O)C
107-87-9
C5H10O
86.13
XNLICIUVMPYHGG-UHFFFAOYSA-N
2-pentanone,methyl propyl ketone,ethyl acetone,pentanone,ethylacetone,propyl methyl ketone,metylopropyloketon,methyl n-propyl ketone,methylpropyl ketone,methyl-propyl-cetone
7895
CHEBI:16472
pentan-2-one
CCCC(=O)C
Viscosity 0.5 mPa.s (20°C)
Refractive Index 1.3890 to 1.3910
Boiling Point 100.0°C to 103.0°C
Water 0.1% Max. (K.F.)
Infrared Spectrum Authentic
Specific Gravity 0.809
Flash Point 7°C
Packaging Glass bottle
Quantity 2.5L
Density 0.8090g/mL
Melting Point -78.0°C
Assay Percent Range 99%
Linear Formula CH3CO(CH2)CH3
Beilstein 01,676
Merck Index 15,6187
Solubility Information Solubility in water: 43g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 86.13
Physical Form Liquid
Percent Purity 99%
Grade Pure
Chemical Name or Material 2-Pentanone
Show More Show Less