Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pentene, cis + trans, 98%

Catalog No. AAA1773622
Click to view available options
Quantity:
25 g
100 g

CAS: 109-68-2 | C5H10 | 70.14 g/mol

2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-68-2
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009384
InChI Key QMMOXUPEWRXHJS-HYXAFXHYSA-N
Synonym trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
PubChem CID 5326161
SMILES CC\C=C/C

Specifications

Density 0.65
Boiling Point 36°C to 38°C
Flash Point −45°C (−49°F)
Odor Petroleum-like
Refractive Index 1.38
Linear Formula CH3CH2CH=CHCH3
Quantity 100 g
UN Number UN3295
Beilstein 969151
Merck Index 14,7123
Solubility Information Not miscible or difficult to mix in water.
IUPAC Name (E)-pent-2-ene
Formula Weight 70.14
Percent Purity 98%
Chemical Name or Material 2-Pentene, cis + trans
Show More Show Less
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 203-695-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.