Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pentenenitrile 90.0+%, TCI America™
SDP

Catalog No. P087025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P087025ML 25 mL
P08705ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P087025ML Supplier TCI America Supplier No. P087025ML

Chemical Identifiers

CAS 13284-42-9
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.12
MDL Number MFCD00001936
InChI Key ISBHMJZRKAFTGE-ONEGZZNKSA-N
Synonym 1-Buten-1-yl Cyanide
PubChem CID 5364360
IUPAC Name (2E)-pent-2-enenitrile
SMILES CC\C=C\C#N

Specifications

Color Yellow
Boiling Point 44°C
Quantity 25 mL
UN Number 1992
Formula Weight 81.12
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pentenenitrile
RTECSNumber SB2256000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.