Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Phenoxyethanol, 94%
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
2-Phenoxyethanol is used as a solvent for cellulose acetate, inks and resins. It is also used as a fixative for perfumes, insect repellents and topical antiseptics. It is utilized as a surface active agent. It acts as an anesthetic in the aquaculture of some fish. Further, it is used as a preservative and cosmetic raw material. In addition to this, it is used in preparation of active pharmaceutical ingredients and dyes.
Solubility
Miscible with water, ethanol, propanol and propylene glycol.
Notes
Incompatible with strong oxidizing agents.
2-Phenoxyethanol is used as a solvent for cellulose acetate, inks and resins. It is also used as a fixative for perfumes, insect repellents and topical antiseptics. It is utilized as a surface active agent. It acts as an anesthetic in the aquaculture of some fish. Further, it is used as a preservative and cosmetic raw material. In addition to this, it is used in preparation of active pharmaceutical ingredients and dyes.
Solubility
Miscible with water, ethanol, propanol and propylene glycol.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 11°C to 14°C |
Density | 1.102 |
Boiling Point | 245°C |
Flash Point | 121°C (249°F) |
Assay Percent Range | 94% |
Refractive Index | 1.537 |
Linear Formula | C6H5O(CH2)2OH |
Quantity | 1000 g |
Beilstein | 1364011 |
Merck Index | 14,7257 |
Show More |
Safety and Handling
Hazard Category | H302-H319 |
Hazard Statement | GHS H Statement H302-H319 Harmful if swallowed. Causes serious eye irritation. |
Precautionary Statement | P264b-P270-P280-P301+P312-P305+P351+P338-P330-P501c |
EINECSNumber | 204-589-7 |
RTECSNumber | KM0350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction