Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxyethanol, 94%

Catalog No. p-7052392
Click to view available options
Quantity:
250 g
1000 g
5000 g

CAS: 122-99-6 | C8H10O2 | 138.166 g/mol

2-Phenoxyethanol is used as a solvent for cellulose acetate, inks and resins. It is also used as a fixative for perfumes, insect repellents and topical antiseptics. It is utilized as a surface active agent. It acts as an anesthetic in the aquaculture of some fish. Further, it is used as a preservative and cosmetic raw material. In addition to this, it is used in preparation of active pharmaceutical ingredients and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenoxyethanol is used as a solvent for cellulose acetate, inks and resins. It is also used as a fixative for perfumes, insect repellents and topical antiseptics. It is utilized as a surface active agent. It acts as an anesthetic in the aquaculture of some fish. Further, it is used as a preservative and cosmetic raw material. In addition to this, it is used in preparation of active pharmaceutical ingredients and dyes.

Solubility
Miscible with water, ethanol, propanol and propylene glycol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-99-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002857
InChI Key QCDWFXQBSFUVSP-UHFFFAOYSA-N
Synonym phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
PubChem CID 31236
ChEBI CHEBI:64275
IUPAC Name 2-phenoxyethanol
SMILES C1=CC=C(C=C1)OCCO

Specifications

Melting Point 11°C to 14°C
Color Colorless
Density 1.102 g/mL
Boiling Point 245°C
Flash Point 121°C (249°F)
Assay Percent Range 94%
Refractive Index 1.537
Linear Formula C6H5O(CH2)2OH
Quantity 250 g
Beilstein 1364011
Merck Index 14,7257
Solubility Information Miscible with water,ethanol,propanol and propylene glycol.
Formula Weight 138.17
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material 2-Phenoxyethanol
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 204-589-7
RTECSNumber KM0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.