Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxytetrahydropyran 90.0+%, TCI America™
SDP

Catalog No. P085325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P085325ML 25 mL
P08535ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P085325ML Supplier TCI America Supplier No. P085325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4203-50-3
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00059801
InChI Key SHRDVLUJLDYXSO-UHFFFAOYNA-N
Synonym Tetrahydro-2-phenoxy-2H-pyran
PubChem CID 107240
IUPAC Name 2-phenoxyoxane
SMILES C1CCC(OC2=CC=CC=C2)OC1

Specifications

Quantity 25 mL
Formula Weight 178.23
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenoxytetrahydropyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.