Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-1,3-indandione 98.0+%, TCI America™
SDP

Catalog No. P102925G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Catalog No. P102925G

Supplier: TCI America P102925G

Chemical Identifiers

CAS 83-12-5
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.243
MDL Number MFCD00003782
InChI Key NFBAXHOPROOJAW-UHFFFAOYSA-N
Synonym phenindione, 2-phenyl-1,3-indandione, dindevan, 2-phenyl-1h-indene-1,3 2h-dione, phenylindanedione, athrombon, diophindane, emandione, fenhydren, phenhydren
PubChem CID 4760
ChEBI CHEBI:8066
IUPAC Name 2-phenylindene-1,3-dione
SMILES C1=CC=C(C=C1)C2C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 222.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenyl-1,3-indandione
RTECSNumber NK6125000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.