Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylacetamide, 99%

Catalog No. aah60626md
Click to view available options
Quantity:
250 mg

CAS: 103-81-1 | C8H9NO | 135.17 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-81-1
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00059193
InChI Key LSBDFXRDZJMBSC-UHFFFAOYSA-N
Synonym benzeneacetamide, phenylacetamide, alpha-toluamide, alpha-phenylacetamide, phenylacetic acid amide, acetamide, 2-phenyl, phenyl-beta-acetylamine, alpha-toluimidic acid, 2-phenyl-acetamide, phenyl acetamide
PubChem CID 7680
ChEBI CHEBI:16562
IUPAC Name 2-phenylacetamide
SMILES NC(=O)CC1=CC=CC=C1

Specifications

Quantity 250 mg
Solubility Information Soluble in methanol.
Formula Weight 135.17
Percent Purity 99%
Chemical Name or Material 2-Phenylacetamide
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 203-147-0
RTECSNumber AC7705000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.