Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylethyl Methyl Ether 98.0+%, TCI America™
SDP

Catalog No. P126325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P126325ML 25 mL
P1263500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P126325ML Supplier TCI America Supplier No. P126325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3558-60-9
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00025912
InChI Key CQLYXIUHVFRXLT-UHFFFAOYSA-N
Synonym Methyl Phenethyl Ether
PubChem CID 19089
IUPAC Name 2-methoxyethylbenzene
SMILES COCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 190°C
Quantity 25 mL
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenylethyl Methyl Ether
EINECSNumber (3)-1035
RTECSNumber KO2230000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.