Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Phenylethyl)phosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. P21221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21221G 1 g
P2122200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21221G Supplier TCI America Supplier No. P21221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4672-30-4
Molecular Formula C8H11O3P
Molecular Weight (g/mol) 186.147
MDL Number MFCD00485102
InChI Key BFDYSJCMAFSRDH-UHFFFAOYSA-N
Synonym Phenethylphosphonic Acid
PubChem CID 78404
IUPAC Name 2-phenylethylphosphonic acid
SMILES C1=CC=C(C=C1)CCP(=O)(O)O

Specifications

Melting Point 138°C
Quantity 1 g
Formula Weight 186.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Phenylethyl)phosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.