Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Phenylethyl Propionate 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
250 g
Specifications
Specifications
Color | Colorless |
Boiling Point | 244°C |
Quantity | 250 g |
Formula Weight | 178.23 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 2-Phenylethyl Propionate |
Safety and Handling
EINECSNumber | (3)-1040&(3)-1045 |
RTECSNumber | UF6488000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction