Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylethyl Propionate 98.0+%, TCI America™
SDP

Catalog No. P182325G
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 122-70-3
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00027008
InChI Key HVGZQCSMLUDISR-UHFFFAOYSA-N
Synonym 2-phenylethyl propionate, phenethyl propionate, propanoic acid, 2-phenylethyl ester, phenylethyl propionate, 2-phenethyl propionate, benzylcarbinyl propionate, ecopco acu, phenethyl alcohol, propionate, phenethyl popanoate, japanese beetle bait
PubChem CID 31225
IUPAC Name 2-phenylethyl propanoate
SMILES CCC(=O)OCCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 244°C
Quantity 25 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenylethyl Propionate
EINECSNumber (3)-1040&(3)-1045
RTECSNumber UF6488000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.