Learn More
2-Phenylimidazole, 98%, Thermo Scientific Chemicals
Description
2-Phenylimidazole is used to prepare complex compounds with ruthenium(III) and also as an intermediate for agrochemical, pharmaceutical and specialty products. It also finds its application as a curing agent for epoxy resins and polyurethane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Phenylimidazole is used to prepare complex compounds with ruthenium(III) and also as an intermediate for agrochemical, pharmaceutical and specialty products. It also finds its application as a curing agent for epoxy resins and polyurethane.
Solubility
Solubility in methanol (almost transparency).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 146°C to 150°C |
Boiling Point | 198°C to 200°C (7 mmHg) |
Flash Point | 200°C (392°F) |
Quantity | 25 g |
Beilstein | 114011 |
Formula Weight | 144.18 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Phenylimidazole |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 211-581-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.