Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylimidazole, 98%

Catalog No. p-7027251
Click to view available options
Quantity:
25 g
100 g

CAS: 670-96-2 | C9H8N2 | 144.18 g/mol

2-Phenylimidazole is used to prepare complex compounds with ruthenium(III) and also as an intermediate for agrochemical, pharmaceutical and specialty products. It also finds its application as a curing agent for epoxy resins and polyurethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylimidazole is used to prepare complex compounds with ruthenium(III) and also as an intermediate for agrochemical, pharmaceutical and specialty products. It also finds its application as a curing agent for epoxy resins and polyurethane.

Solubility
Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 670-96-2
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00005186
InChI Key ZCUJYXPAKHMBAZ-UHFFFAOYSA-N
Synonym 2-phenylimidazole, 1h-imidazole, 2-phenyl, imidazole, 2-phenyl, 2-phenyl imidazole, 2-phenyl-1h-imidazol, unii-7r2590hm4l, chembl14189, phenyl imidazole, 2-phenyl-imidazole, piy
PubChem CID 69591
IUPAC Name 2-phenyl-1H-imidazole
SMILES N1C=CN=C1C1=CC=CC=C1

Specifications

Melting Point 146°C to 150°C
Boiling Point 198°C to 200°C (7 mmHg)
Flash Point 200°C (392°F)
Quantity 25 g
Beilstein 114011
Formula Weight 144.18
Percent Purity ≥98%
Chemical Name or Material 2-Phenylimidazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-581-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.