Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylimidazole, 98%

Catalog No. p-182571
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 670-96-2 | C9H8N2 | 144.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 670-96-2
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00005186
InChI Key ZCUJYXPAKHMBAZ-UHFFFAOYSA-N
Synonym 2-phenylimidazole, 1h-imidazole, 2-phenyl, imidazole, 2-phenyl, 2-phenyl imidazole, 2-phenyl-1h-imidazol, unii-7r2590hm4l, chembl14189, phenyl imidazole, 2-phenyl-imidazole, piy
PubChem CID 69591
IUPAC Name 2-phenyl-1H-imidazole
SMILES N1C=CN=C1C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 141.0°C to 148.0°C
Color Yellow to Beige
Boiling Point 340.0°C
Flash Point 200°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 23, 182
Solubility Information Solubility in water: insoluble
Formula Weight 144.18
Percent Purity 98%
Physical Form Crystalline Powder, Powder or Granular
Chemical Name or Material 2-Phenylimidazole, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse
EINECSNumber 211-581-7
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.